Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1064 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001076.1 | NP_001001076.1 | 1 | 647 |
Gene Symbol: LOC100910928 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249611.1 | XP_006249673.1 | 1 | 647 |