Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRIG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001312686.1 | NP_001299615.1 | 11 | 1540 | ||
NM_014813.2 | NP_055628.1 | 10 | 1428 | |||
XM_005271369.2 | XP_005271426.1 | 10 | 1428 | |||
XM_017002952.1 | XP_016858441.1 | 9 | 952 | |||
GenBank mRNA | AB018349.1 | - | 10 | 1389 | ||
AK291031.1 | - | 10 | 1420 | |||
BC117368.1 | - | 10 | 1252 | 40125919 | ||
BC117370.1 | - | 10 | 1252 | 40125921 | ||
CR936669.1 | - | 11 | 1531 |