Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271779.1 | NP_001258708.1 | 7 | 1194 | ||
NM_001271780.1 | NP_001258709.1 | 6 | 909 | |||
NM_001271781.1 | NP_001258710.1 | 6 | 784 | |||
NM_014814.2 | NP_055629.1 | 6 | 1035 | |||
XM_005265619.1 | XP_005265676.1 | 6 | 1131 | |||
XM_017007569.1 | XP_016863058.1 | 7 | 937 | |||
GenBank mRNA | AF215935.1 | - | 6 | 901 | ||
AF530062.1 | - | 6 | 848 | |||
AK055210.1 | - | 2 | 1537 | |||
AK095059.1 | - | 5 | 1617 | |||
AK290205.1 | - | 6 | 889 | |||
AL529900.3 | - | 6 | 770 | |||
AL832109.1 | - | 1 | 4145 | |||
AY359879.1 | - | 6 | 881 | |||
AY568086.1 | - | 6 | 873 | |||
BC000630.1 | - | 6 | 865 | 3344973 | ||
BC000904.2 | - | 6 | 886 | 2900971 | ||
BC012369.1 | - | 6 | 883 | 3858094 | ||
BX647742.1 | - | 7 | 1061 | |||
D14663.1 | - | 6 | 873 |