Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CDC42BPA SIRNA Assays ›
Gene Symbol:
CDC42BPA
Gene Name:
CDC42 binding protein kinase alpha
Gene Aliases:
MRCK, MRCKA, PK428
Chromosome Location:
Chr.1: 226989865 - 227318492 on Build GRCh38
UniGene ID:
Hs.35433
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
22258
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CDC42BPA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_003607.3
NP_003598.2
28
4654
NM_014826.4
NP_055641.3
27
4411
XM_005273317.3
XP_005273374.1
30
4919
XM_005273318.3
XP_005273375.1
30
4910
XM_005273320.3
XP_005273377.1
29
4901
XM_005273321.3
XP_005273378.1
28
4766
XM_005273322.3
XP_005273379.1
29
4871
XM_005273324.3
XP_005273381.1
29
4901
XM_006711834.3
XP_006711897.1
29
4871
XM_006711835.3
XP_006711898.1
29
4814
XM_011544306.2
XP_011542608.1
31
5213
XM_011544307.2
XP_011542609.1
30
4853
XM_011544308.2
XP_011542610.1
31
4958
XM_011544309.2
XP_011542611.1
30
4715
XM_017002578.1
XP_016858067.1
30
4910
XM_017002579.1
XP_016858068.1
32
5371
XM_017002580.1
XP_016858069.1
30
3658
XM_017002581.1
XP_016858070.1
30
5284
XM_017002582.1
XP_016858071.1
28
4766
GenBank mRNA
AB007920.2
-
12
1318
AB208968.1
-
27
4285
AJ518975.1
-
28
3711
AJ518976.1
-
27
3468
AK027000.1
-
4
403
AY166586.1
-
27
3468
BC136333.1
-
28
3678
9020320
BX648219.1
-
6
635
BX648693.1
-
15
1696
CR933723.1
-
15
1696
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
cell migration
actomyosin structure organization
actin cytoskeleton reorganization
intracellular signal transduction
magnesium ion binding
protein serine/threonine kinase activity
protein binding
ATP binding
identical protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-sd57s:80/100.66.78.144:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline