Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001718.1 | NP_001001718.1 | 7 | 979 | ||
XM_006246639.2 | XP_006246701.1 | 8 | 1179 | |||
XM_006246640.1 | XP_006246702.1 | 6 | 488 | |||
GenBank mRNA | AY623900.1 | - | 7 | 979 | ||
BC158569.1 | - | 8 | 1182 | 9101477 |