Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286658.2 | NP_001273587.1 | 20 | 5439 | ||
NM_001286659.2 | NP_001273588.1 | 19 | 5274 | |||
NM_014832.4 | NP_055647.2 | 21 | 5463 | |||
XM_005266603.1 | XP_005266660.1 | 20 | 5866 | |||
XM_005266605.2 | XP_005266662.1 | 21 | 4791 | |||
XM_006719903.3 | XP_006719966.1 | 21 | 5151 | |||
XM_011535331.1 | XP_011533633.1 | 20 | 5830 | |||
XM_017020882.1 | XP_016876371.1 | 19 | 4483 | |||
XM_017020883.1 | XP_016876372.1 | 18 | 4477 | |||
XM_017020884.1 | XP_016876373.1 | 9 | 4049 | |||
GenBank mRNA | AB011175.1 | - | 22 | 5412 | ||
BC151239.1 | - | 22 | 5404 | 8860512 | ||
CR749622.1 | - | 11 | 3243 |