Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304360.1 | NP_001291289.1 | 36 | 4595 | ||
XM_011542333.2 | XP_011540635.2 | 36 | 4512 | |||
XM_011542335.1 | XP_011540637.1 | 31 | 4024 | |||
XM_011542336.2 | XP_011540638.1 | 31 | 5759 | |||
XM_017002641.1 | XP_016858130.1 | 37 | 4721 | |||
XM_017002642.1 | XP_016858131.1 | 37 | 4721 | |||
XM_017002643.1 | XP_016858132.1 | 37 | 4721 | |||
XM_017002644.1 | XP_016858133.1 | 17 | 2139 | |||
GenBank mRNA | AL137547.1 | - | 7 | 1945 | ||
BC114926.1 | - | 8 | 1287 | 40026076 |