Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303279.1 | NP_001290208.1 | 14 | 1957 | ||
NM_015194.2 | NP_056009.1 | 14 | 1957 | |||
XM_017024685.1 | XP_016880174.1 | 14 | 1928 | |||
GenBank mRNA | AB018270.2 | - | 14 | 1668 | ||
AK026920.1 | - | 11 | 1253 | |||
AK127942.1 | - | 14 | 1928 | |||
BC146763.1 | - | 14 | 1668 | 9007203 |