Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GAPVD1 SIRNA Assays ›
Gene Symbol:
GAPVD1
Gene Name:
GTPase activating protein and VPS9 domains 1
Gene Aliases:
GAPEX5, GAPex-5, RAP6
Chromosome Location:
Chr.9: 125261794 - 125365011 on Build GRCh38
UniGene ID:
Hs.495134
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
222688
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GAPVD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282679.1
NP_001269608.1
27
4517
NM_001282680.1
NP_001269609.1
27
4463
NM_001282681.1
NP_001269610.1
24
4068
NM_015635.3
NP_056450.2
26
4378
XM_005251901.3
XP_005251958.1
26
4327
XM_005251904.3
XP_005251961.1
25
4243
XM_006717044.3
XP_006717107.1
26
4249
XM_011518499.2
XP_011516801.1
27
4422
XM_011518500.2
XP_011516802.1
28
4539
XM_011518502.2
XP_011516804.1
27
4350
XM_011518506.2
XP_011516808.1
27
4347
XM_011518507.2
XP_011516809.1
26
4346
XM_017014600.1
XP_016870089.1
26
4232
XM_017014601.1
XP_016870090.1
25
4254
XM_017014602.1
XP_016870091.1
27
4474
XM_017014603.1
XP_016870092.1
27
4421
XM_017014604.1
XP_016870093.1
28
4523
XM_017014605.1
XP_016870094.1
25
4254
XM_017014606.1
XP_016870095.1
27
4368
XM_017014607.1
XP_016870096.1
27
4377
XM_017014608.1
XP_016870097.1
25
4187
XM_017014609.1
XP_016870098.1
26
4355
XM_017014610.1
XP_016870099.1
24
4173
XM_017014611.1
XP_016870100.1
25
4166
GenBank mRNA
AB040954.3
-
27
4443
AK021533.1
-
5
1769
AK023841.1
-
18
2412
AK096224.1
-
8
1982
AL080196.1
-
16
2160
AL834325.1
-
6
708
BC013635.1
-
10
1368
BC114937.1
-
27
4346
40083103
BC114962.1
-
24
4101
40083104
DQ233254.1
-
24
4036
Back To Top
More Information
Panther Classification:
Molecular Function -
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
signal transduction
positive regulation of GTPase activity
regulation of protein transport
cell-cell adhesion
guanyl-nucleotide exchange factor activity
GTPase activator activity
GTPase activating protein binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline