Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017554.2 | NP_060024.2 | 6 | 1711 | ||
XM_011512928.2 | XP_011511230.1 | 6 | 1802 | |||
XM_011512929.2 | XP_011511231.1 | 6 | 1810 | |||
GenBank mRNA | AK098816.1 | - | 1 | 624 | ||
AK304269.1 | - | 6 | 1679 | |||
AY134858.1 | - | 1 | 884 | |||
BX648758.1 | - | 2 | 1030 | |||
DQ063584.1 | - | 3 | 1175 | |||
DQ063585.1 | - | 3 | 1175 |