Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014865.3 | NP_055680.3 | 26, 27 | 3531 | ||
GenBank mRNA | AK128354.1 | - | 26, 27 | 3443 | ||
BC028182.1 | - | 26, 27 | 3543 | 5395775 | ||
D63880.1 | - | 26, 27 | 4284 |