Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HELZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014877.3 | NP_055692.2 | 30, 31 | 4919 | ||
XM_005257888.4 | XP_005257945.1 | 31, 32 | 5048 | |||
XM_005257889.4 | XP_005257946.1 | 30, 31 | 4970 | |||
XM_006722214.3 | XP_006722277.1 | 31, 32 | 5051 | |||
XM_006722215.3 | XP_006722278.1 | 29, 30 | 4923 | |||
XM_006722216.3 | XP_006722279.1 | 20, 21 | 3753 | |||
XM_011525544.2 | XP_011523846.1 | 31, 32 | 5122 | |||
XM_017025477.1 | XP_016880966.1 | 28, 29 | 4839 | |||
XM_017025478.1 | XP_016880967.1 | 31, 32 | 5104 | |||
GenBank mRNA | BC094881.1 | - | 28, 29 | 4945 | 3076113 | |
BC130582.1 | - | 30, 31 | 4899 | 40146613 | ||
BC144083.1 | - | 30, 31 | 4902 | 9052603 | ||
D29677.2 | - | 29, 30 | 4877 |