Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD302 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198764.1 | NP_001185693.1 | 3 | 360 | ||
NM_014880.4 | NP_055695.2 | 4 | 471 | |||
GenBank mRNA | AF499000.1 | - | 4 | 425 | ||
AY314007.1 | - | 4 | 458 | |||
BC020646.1 | - | 3 | 307 | 4716449 | ||
D14664.1 | - | 4 | 412 | |||
DB217404.1 | - | 4 | 471 | |||
DB460994.1 | - | 3 | 327 |
Gene Symbol: LY75-CD302 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198759.1 | NP_001185688.1 | 37 | 5411 | ||
NM_001198760.1 | NP_001185689.1 | 36 | 5243 | |||
GenBank mRNA | AY184222.1 | - | 37 | 5337 | ||
AY314006.1 | - | 36 | 5169 |