Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256706.1 | NP_001243635.1 | 5 | 762 | ||
NM_001256707.1 | NP_001243636.1 | 6 | 835 | |||
NM_001256708.1 | NP_001243637.1 | 5 | 968 | |||
NM_001256709.1 | NP_001243638.1 | 7 | 903 | |||
NM_001256710.1 | NP_001243639.1 | 6 | 800 | |||
NM_001256711.1 | NP_001243640.1 | 7 | 873 | |||
NM_001256712.1 | NP_001243641.1 | 4 | 630 | |||
NM_001318367.1 | NP_001305296.1 | 5 | 800 | |||
NM_014885.4 | NP_055700.2 | 5 | 751 | |||
NR_046345.1 | 3 | 514 | ||||
XM_011531524.2 | XP_011529826.1 | 6 | 991 | |||
XM_011531531.2 | XP_011529833.1 | 4 | 657 | |||
XM_017007638.1 | XP_016863127.1 | 5 | 778 | |||
XM_017007639.1 | XP_016863128.1 | 6 | 1427 | |||
XM_017007640.1 | XP_016863129.1 | 5 | 819 | |||
XM_017007643.1 | XP_016863132.1 | 6 | 816 | |||
XM_017007644.1 | XP_016863133.1 | 6 | 790 | |||
GenBank mRNA | AB012109.1 | - | 5 | 749 | ||
AF132794.1 | - | 5 | 706 | |||
AL080090.1 | - | 6 | 809 | |||
BC005217.1 | - | 5 | 728 | 3929766 | ||
BI488888.1 | - | 3 | 514 |