Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PITRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242307.1 | NP_001229236.1 | 27 | 3360 | ||
NM_001242309.1 | NP_001229238.1 | 24 | 3056 | |||
NM_014889.3 | NP_055704.2 | 27 | 3357 | |||
XM_005252345.3 | XP_005252402.1 | 25 | 3159 | |||
XM_017015470.1 | XP_016870959.1 | 27 | 3425 | |||
XM_017015471.1 | XP_016870960.1 | 25 | 3227 | |||
XM_017015472.1 | XP_016870961.1 | 27 | 3254 | |||
XM_017015473.1 | XP_016870962.1 | 25 | 3056 | |||
GenBank mRNA | AB029027.2 | - | 17 | 2086 | ||
AF061243.1 | - | 27 | 3304 | |||
AK001183.1 | - | 12 | 1543 | |||
AK002061.1 | - | 27 | 3325 | |||
AK023476.1 | - | 20 | 2472 | |||
AK225084.1 | - | 27 | 3295 | |||
AK299464.1 | - | 26 | 3103 | |||
AL117409.1 | - | 9 | 3110 | |||
BC001150.1 | - | 21 | 2657 | 3503609 | ||
BC005025.1 | - | 27 | 3292 | 4040225 | ||
BC045577.1 | - | 25 | 3569 | |||
BC095422.1 | - | 27 | 3298 | 4822631 | ||
BX649017.1 | - | 25 | 3975 | |||
CR749279.1 | - | 21 | 5508 |