Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1587 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000913.1 | NP_001000913.1 | 1 | 552 | ||
XM_008769791.1 | XP_008768013.1 | 2 | 3522 | |||
XM_008769792.1 | XP_008768014.1 | 3 | 3264 | |||
XM_008769793.1 | XP_008768015.1 | 3 | 3265 |