Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF652 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145365.1 | NP_001138837.1 | 6 | 2600 | ||
NM_014897.2 | NP_055712.1 | 6 | 2455 | |||
XM_005257166.2 | XP_005257223.1 | 5 | 1807 | |||
GenBank mRNA | AB023141.1 | - | 6 | 2455 | ||
AK292667.1 | - | 6 | 2590 | |||
BC152440.1 | - | 6 | 2455 | 8862563 | ||
CR933680.1 | - | 6 | 2451 |