Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hexdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001333.2 | NP_001001333.2 | 5 | 853 | ||
NM_001146073.1 | NP_001139545.1 | 4 | 439 | |||
GenBank mRNA | AK154237.1 | - | 5 | 827 | ||
BC056487.1 | - | 5 | 2434 | |||
BC069960.1 | - | 4 | 391 | 5007884 | ||
EF437388.1 | - | 1 | 12 | |||
FM204885.1 | - | 4 | 390 | |||
FM204886.1 | - | 4 | 390 |