Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tas2r138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001451.1 | NP_001001451.1 | 1 | 888 | ||
GenBank mRNA | AY916509.1 | - | 1 | 888 | ||
BC128016.1 | - | 1 | 892 | 40136088 |