Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001495.2 | NP_001001495.2 | 8 | 3605 | ||
GenBank mRNA | AK033534.1 | - | 8 | 3413 |