Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP27C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001665.3 | NP_001001665.3 | 6 | 904 | ||
XM_017003960.1 | XP_016859449.1 | 7 | 1094 | |||
XM_017003961.1 | XP_016859450.1 | 5 | 795 | |||
GenBank mRNA | AK131190.1 | - | 6 | 885 | ||
BC039307.1 | - | 6 | 904 | 5266370 |