Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA31D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145197.1 | NP_001138669.1 | 4 | 2364 | ||
GenBank mRNA | AK125847.1 | - | 4 | 2309 | ||
AK308327.1 | - | 4 | 2305 |
Gene Symbol: SPATA31D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001670.2 | NP_001001670.1 | 4 | 2334 | ||
XM_017014710.1 | XP_016870199.1 | 4 | 2312 | |||
GenBank mRNA | AK128831.1 | - | 4 | 2334 |
Gene Symbol: SPATA31D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207416.2 | NP_997299.2 | 4 | 2364 | ||
GenBank mRNA | AK126070.1 | - | 1 | 1067 |
Gene Symbol: SPATA31D5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026851.1 | 4 | 2330 | |||
GenBank mRNA | AK125938.1 | - | 4 | 2214 | ||
AK128229.1 | - | 5 | 2539 |