Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sohlh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001714.1 | NP_001001714.1 | 4 | 468 | ||
GenBank mRNA | AY623913.1 | - | 4 | 468 | ||
BC139189.1 | - | 4 | 436 | 8862211 | ||
BC139190.1 | - | 4 | 436 | 8862212 |