Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CC2D2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001732.3 | NP_001001732.2 | 9 | 1712 | ||
NM_001159747.1 | NP_001153219.1 | 12 | 1982 | |||
XM_011539789.2 | XP_011538091.1 | 19 | 3043 | |||
XM_011539790.2 | XP_011538092.1 | 18 | 2929 | |||
XM_011539791.2 | XP_011538093.1 | 18 | 2860 | |||
XM_011539792.2 | XP_011538094.1 | 13 | 2165 | |||
XM_011539793.2 | XP_011538095.1 | 12 | 3144 | |||
XM_011539795.2 | XP_011538097.1 | 11 | 2961 | |||
XM_017016240.1 | XP_016871729.1 | 12 | 2221 | |||
GenBank mRNA | BC075861.1 | - | 9 | 1677 | 5171414 |