Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itprip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001738.2 | NP_001001738.2 | 2 | 1849 | ||
GenBank mRNA | AK160686.1 | - | 2 | 1851 | ||
AK165192.1 | - | 2 | 1828 | |||
AK171522.1 | - | 2 | 1755 | |||
AK171539.1 | - | 2 | 1755 | |||
BC063749.1 | - | 3 | 1815 | 6813779 |