Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004686.4 | NP_004677.3 | 14 | 2404 | ||
XM_006716414.3 | XP_006716477.2 | 9 | 2770 | |||
XM_011544692.2 | XP_011542994.1 | 11 | 1843 | |||
XM_011544693.2 | XP_011542995.1 | 10 | 2305 | |||
XM_017013956.1 | XP_016869445.1 | 13 | 2093 | |||
GenBank mRNA | AK294468.1 | - | 14 | 2165 | ||
AK303762.1 | - | 6 | 1227 | |||
AK303803.1 | - | 5 | 1175 | |||
AK316022.1 | - | 14 | 2214 | |||
AK316283.1 | - | 14 | 2224 | |||
BC130577.1 | - | 14 | 2190 | 40146608 | ||
BC130579.1 | - | 14 | 2190 | 40146610 | ||
CR749240.1 | - | 14 | 2202 |