Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SALL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291446.1 | NP_001278375.1 | 4 | 3877 | ||
NM_001291447.1 | NP_001278376.1 | 4 | 3857 | |||
NM_005407.2 | NP_005398.2 | 2 | 4870 | |||
NR_111966.1 | 3 | 1778 | ||||
NR_111967.1 | 3 | 1758 | ||||
XM_011537064.1 | XP_011535366.1 | 3 | 4208 | |||
XM_011537065.1 | XP_011535367.1 | 3 | 4354 | |||
GenBank mRNA | AB002358.2 | - | 2 | 4840 | ||
AK001276.1 | - | 3 | 1494 | |||
BC090958.1 | - | 3 | 1578 | |||
X98834.1 | - | 2 | 4755 |