Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ephb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290753.1 | NP_001277682.1 | 16 | 4138 | ||
NM_010142.3 | NP_034272.1 | 16 | 4135 | |||
GenBank mRNA | BC043088.1 | - | 16 | 4059 | 6418088 | |
BC062924.1 | - | 16 | 4083 |