Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ryk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042607.1 | NP_001036072.1 | 15 | 2048 | ||
NM_001284258.1 | NP_001271187.1 | 14 | 1926 | |||
NM_013649.3 | NP_038677.3 | 15 | 2057 | |||
GenBank mRNA | AK076449.1 | - | 9 | 1147 | ||
AK144355.1 | - | 14 | 1866 | |||
BC006963.1 | - | 11 | 1356 | |||
BC032275.1 | - | 15 | 1782 | 5370100 | ||
L02210.1 | - | 15 | 2032 | |||
L21707.1 | - | 15 | 2066 |