Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014935.4 | NP_055750.2 | 6 | 626 | ||
XM_005244966.4 | XP_005245023.2 | 6 | 493 | |||
XM_005244967.4 | XP_005245024.2 | 6 | 492 | |||
XM_005244968.4 | XP_005245025.2 | 6 | 492 | |||
XM_006711212.3 | XP_006711275.1 | 6 | 719 | |||
XM_006711213.3 | XP_006711276.2 | 6 | 493 | |||
XM_006711214.3 | XP_006711277.2 | 6 | 493 | |||
XM_006711215.3 | XP_006711278.2 | 6 | 493 | |||
XM_006711217.3 | XP_006711280.2 | 6 | 493 | |||
XM_006711219.3 | XP_006711282.2 | 6 | 493 | |||
XM_006711221.2 | XP_006711284.1 | 6 | 817 | |||
XM_006711222.3 | XP_006711285.1 | 6 | 802 | |||
XM_011509295.2 | XP_011507597.1 | 6 | 493 | |||
XM_011509296.2 | XP_011507598.1 | 6 | 493 | |||
XM_011509297.2 | XP_011507599.1 | 8 | 1319 | |||
XM_011509298.2 | XP_011507600.1 | 6 | 602 | |||
XM_011509299.2 | XP_011507601.2 | 6 | 608 | |||
XM_011509301.2 | XP_011507603.1 | 6 | 4498 | |||
XM_011509304.2 | XP_011507606.1 | 6 | 447 | |||
XM_017000687.1 | XP_016856176.1 | 6 | 493 | |||
XM_017000688.1 | XP_016856177.1 | 6 | 493 | |||
XM_017000689.1 | XP_016856178.1 | 6 | 493 | |||
XM_017000690.1 | XP_016856179.1 | 6 | 4498 | |||
GenBank mRNA | AB023186.1 | - | 6 | 613 | ||
BC152475.1 | - | 6 | 613 | 8862612 |