Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNIK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161560.2 | NP_001155032.1 | 17 | 2329 | ||
NM_001161561.2 | NP_001155033.1 | 16 | 2242 | |||
NM_001161562.2 | NP_001155034.1 | 16 | 2242 | |||
NM_001161563.2 | NP_001155035.1 | 16 | 2164 | |||
NM_001161564.2 | NP_001155036.1 | 16 | 2164 | |||
NM_001161565.2 | NP_001155037.1 | 15 | 2077 | |||
NM_001161566.2 | NP_001155038.1 | 15 | 2077 | |||
NM_015028.3 | NP_055843.1 | 17 | 2329 | |||
GenBank mRNA | AB011123.2 | - | 17 | 2250 | ||
AF172264.1 | - | 17 | 1984 | |||
AF172265.1 | - | 16 | 1897 | |||
AF172266.1 | - | 16 | 1819 | |||
AF172267.1 | - | 17 | 1984 | |||
AF172268.1 | - | 15 | 1732 | |||
AF172269.1 | - | 16 | 1897 | |||
AF172270.1 | - | 16 | 1819 | |||
AF172271.1 | - | 15 | 1732 | |||
AK291056.1 | - | 15 | 2077 | |||
BC150256.1 | - | 17 | 2250 | 9007184 |