Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128431.2 | NP_001121903.1 | 5 | 825 | ||
NM_001135153.1 | NP_001128625.1 | 5 | 866 | |||
NM_001135154.1 | NP_001128626.1 | 5 | 825 | |||
NM_015359.4 | NP_056174.2 | 5 | 825 | |||
XM_005273465.2 | XP_005273522.1 | 6 | 913 | |||
XM_005273466.4 | XP_005273523.1 | 5 | 897 | |||
XM_006716323.2 | XP_006716386.1 | 7 | 1111 | |||
XM_006716324.2 | XP_006716387.1 | 6 | 902 | |||
XM_011544478.2 | XP_011542780.1 | 6 | 977 | |||
XM_017013292.1 | XP_016868781.1 | 6 | 993 | |||
XM_017013293.1 | XP_016868782.1 | 7 | 1111 | |||
XM_017013294.1 | XP_016868783.1 | 7 | 1020 | |||
XM_017013295.1 | XP_016868784.1 | 5 | 1030 | |||
GenBank mRNA | AK172810.1 | - | 5 | 796 | ||
AK295807.1 | - | 5 | 741 | |||
BC015770.1 | - | 5 | 787 | 4865469 | ||
D31887.1 | - | 5 | 769 | |||
KU178568.1 | - | 4 | 650 | |||
KU178569.1 | - | 4 | 650 | |||
KU178570.1 | - | 4 | 650 | |||
KU178571.1 | - | 4 | 650 |