Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128431.2 | NP_001121903.1 | 9 | 4474 | ||
NM_001135153.1 | NP_001128625.1 | 9 | 4515 | |||
NM_015359.4 | NP_056174.2 | 9 | 4474 | |||
XM_005273465.2 | XP_005273522.1 | 10 | 4562 | |||
XM_005273466.4 | XP_005273523.1 | 9 | 4546 | |||
XM_006716323.2 | XP_006716386.1 | 11 | 4760 | |||
XM_006716324.2 | XP_006716387.1 | 10 | 4551 | |||
XM_011544478.2 | XP_011542780.1 | 10 | 4626 | |||
XM_017013292.1 | XP_016868781.1 | 10 | 4642 | |||
XM_017013293.1 | XP_016868782.1 | 11 | 4760 | |||
XM_017013294.1 | XP_016868783.1 | 11 | 4669 | |||
XM_017013295.1 | XP_016868784.1 | 9 | 4679 | |||
GenBank mRNA | AK172810.1 | - | 9 | 4444 | ||
BX648538.1 | - | 2 | 2828 | |||
D31887.1 | - | 9 | 4418 |