Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNPLA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256007.2 | NP_001242936.1 | 3 | 800 | ||
NM_001256008.2 | NP_001242937.1 | 2 | 754 | |||
NM_001256009.2 | NP_001242938.1 | 2 | 754 | |||
NM_001256010.2 | NP_001242939.1 | 2 | 627 | |||
NM_001256011.2 | NP_001242940.1 | 3 | 488 | |||
NM_015723.4 | NP_056538.1 | 4 | 912 | |||
XM_005250396.4 | XP_005250453.1 | 3 | 1756 | |||
XM_011516274.2 | XP_011514576.1 | 3 | 1629 | |||
XM_011516275.2 | XP_011514577.1 | 3 | 1629 | |||
XM_017012261.1 | XP_016867750.1 | 2 | 1583 | |||
GenBank mRNA | AB041261.1 | - | 2 | 580 | ||
AF263613.1 | - | 3 | 606 | |||
AK024335.1 | - | 3 | 488 | |||
AL834147.1 | - | 2 | 507 | |||
BC032999.2 | - | 4 | 788 | 4829433 | ||
BX647865.1 | - | 2 | 473 |