Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNPLA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256007.2 | NP_001242936.1 | 10 | 2436 | ||
NM_001256008.2 | NP_001242937.1 | 9 | 2390 | |||
NM_001256009.2 | NP_001242938.1 | 9 | 2204 | |||
NM_001256010.2 | NP_001242939.1 | 9 | 2263 | |||
NM_001256011.2 | NP_001242940.1 | 10 | 2124 | |||
NM_015723.4 | NP_056538.1 | 11 | 2548 | |||
XM_005250396.4 | XP_005250453.1 | 10 | 3206 | |||
XM_011516274.2 | XP_011514576.1 | 10 | 3265 | |||
XM_011516275.2 | XP_011514577.1 | 10 | 3079 | |||
XM_017012261.1 | XP_016867750.1 | 9 | 3033 | |||
GenBank mRNA | AB041261.1 | - | 9 | 2216 | ||
AF217519.1 | - | 2 | 329 | |||
AF263613.1 | - | 10 | 2242 | |||
AK002115.1 | - | 6 | 736 | |||
AK024335.1 | - | 10 | 2124 | |||
AK291755.1 | - | 10 | 2253 | |||
AL834147.1 | - | 9 | 1957 | |||
BC005203.1 | - | 6 | 702 | |||
BC032999.2 | - | 11 | 2424 | 4829433 | ||
BX647865.1 | - | 9 | 2109 |