Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015793.2 | NP_056608.1 | 804 | |||
GenBank mRNA | AK087709.1 | - | 804 | |||
AK135682.1 | - | 800 | ||||
BC138018.1 | - | 798 | 8861037 |
Gene Symbol: Fbxw28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177419.1 | NP_001170890.1 | 6 | 804 | ||
NM_001177420.1 | NP_001170891.1 | 6 | 804 | |||
GenBank mRNA | AK143322.1 | - | 6 | 800 | ||
AK162190.1 | - | 6 | 802 | |||
BC084592.1 | - | 6 | 795 |