Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015793.2 | NP_056608.1 | 483 | |||
GenBank mRNA | AK087709.1 | - | 483 | |||
AK135682.1 | - | 479 | ||||
AK135771.1 | - | 477 | ||||
AK166611.1 | - | 480 | ||||
AK166612.1 | - | 480 | ||||
BC099372.1 | - | 467 | 30938910 | |||
BC138018.1 | - | 477 | 8861037 | |||
BC145098.1 | - | 473 | 9053646 |
Gene Symbol: Fbxw28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177419.1 | NP_001170890.1 | 5 | 483 | ||
NM_001177420.1 | NP_001170891.1 | 5 | 483 | |||
GenBank mRNA | AK143322.1 | - | 5 | 479 | ||
AK162190.1 | - | 5 | 481 | |||
BC084592.1 | - | 5 | 474 |