Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPLAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017570.4 | NP_060040.1 | 18 | 2557 | ||
XM_011516960.1 | XP_011515262.1 | 18 | 2833 | |||
GenBank mRNA | AB122018.1 | - | 17 | 2453 | ||
AF217994.2 | - | 105 | ||||
AL096750.1 | - | 10 | 1542 | |||
BC036944.1 | - | 13 | 1744 | |||
BC041576.1 | - | 3 | 368 | |||
BC142672.1 | - | 18 | 2557 | 40148181 |