Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppfibp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170433.1 | NP_001163904.1 | 28 | 3342 | ||
NM_026221.2 | NP_080497.1 | 27 | 3309 | |||
GenBank mRNA | AK014559.1 | - | 11 | 1475 | ||
AK146575.1 | - | 27 | 3293 | |||
AK154482.1 | - | 27 | 3254 | |||
AK173133.1 | - | 28 | 3247 | |||
BC035209.1 | - | 27 | 3155 | |||
BC049862.1 | - | 6 | 803 | |||
BC058176.1 | - | 15 | 1845 | |||
BC060078.1 | - | 18 | 3582 | |||
BC071257.1 | - | 8 | 1017 | |||
BC072603.1 | - | 27 | 3286 | 6830850 |