Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2610034B18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027420.4 | NP_081696.1 | 6 | 1941 | ||
GenBank mRNA | AK051261.1 | - | 6 | 1942 |