Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glipr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027450.3 | NP_081726.1 | 5 | 730 | ||
GenBank mRNA | AK017557.1 | - | 5 | 707 | ||
AK141459.1 | - | 5 | 721 | |||
AK154433.1 | - | 5 | 708 | |||
AK167595.1 | - | 5 | 730 | |||
BC031750.1 | - | 5 | 730 | 4037002 |