Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027973.3 | NP_082249.1 | 8 | 748 | ||
GenBank mRNA | AK006479.1 | - | 8 | 757 | ||
AK050386.1 | - | 8 | 749 | |||
AK081703.1 | - | 8 | 736 | |||
AY366362.1 | - | 4 | 280 | |||
BC094675.1 | - | 9 | 868 | |||
BC117784.2 | - | 8 | 749 | 40092112 | ||
BC117785.2 | - | 8 | 749 | 40092113 |