Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028376.3 | NP_082652.1 | 1, 2 | 975 | ||
GenBank mRNA | AK013595.1 | - | 1, 2 | 977 | ||
AY495949.1 | - | 2, 3 | 518 | |||
BC094614.1 | - | 1, 2 | 1248 | |||
BC116888.1 | - | 1, 2 | 819 | 40126207 | ||
BC116890.1 | - | 2, 3 | 406 | 40126209 |