Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acacb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133904.2 | NP_598665.2 | 14 | 2344 | ||
GenBank mRNA | AK147463.1 | - | 3 | 539 | ||
AY451394.1 | - | 14 | 2344 |