Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr873 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146561.1 | NP_666772.1 | 1 | 53 | ||
GenBank mRNA | BC139140.1 | - | 1 | 136 | 8862162 |