Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010307.3 | NP_034437.1 | 4 | 560 | ||
NM_177137.5 | NP_796111.2 | 4 | 725 | |||
GenBank mRNA | AK032407.1 | - | 4 | 371 | ||
AK035952.1 | - | 4 | 726 | |||
AK045484.1 | - | 4 | 690 | |||
AK158831.1 | - | 4 | 561 | |||
AY179169.1 | - | 4 | 294 | |||
BC078439.1 | - | 4 | 687 | 5685967 |