Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPH3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143854.1 | NP_001137326.1 | 5 | 557 | ||
NM_014954.3 | NP_055769.2 | 4 | 545 | |||
XM_011538048.1 | XP_011536350.1 | 5 | 361 | |||
GenBank mRNA | AB023202.1 | - | 5 | 478 | ||
AK295696.1 | - | 5 | 511 | |||
AK316112.1 | - | 5 | 392 | |||
BC002605.2 | - | 3 | 283 | |||
BC009843.2 | - | 3 | 206 | |||
BC017259.2 | - | 4 | 352 | 3510158 | ||
DA332963.1 | - | 5 | 557 |