Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam71f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289663.1 | NP_001276592.1 | 1 | 236 | ||
NM_001289664.1 | NP_001276593.1 | 1 | 236 | |||
NM_001289665.1 | NP_001276594.1 | 1 | 236 | |||
NM_207258.3 | NP_997141.2 | 1 | 236 | |||
GenBank mRNA | AB473725.1 | - | 1 | 177 | ||
AK133908.1 | - | 1 | 237 | |||
BC048651.1 | - | 1 | 237 | |||
BC147513.1 | - | 1 | 219 | 9056505 | ||
BC172074.1 | - | 1 | 219 | 9054360 | ||
BC172085.1 | - | 1 | 219 | |||
BC172086.1 | - | 1 | 219 | 9054372 |