Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnot6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290741.1 | NP_001277670.1 | 2, 3 | 309 | ||
NM_212484.2 | NP_997649.1 | 3, 4 | 678 | |||
GenBank mRNA | AK181033.1 | - | 1, 2 | 39 | ||
AK184045.1 | - | 1, 2 | 39 | |||
AK217278.1 | - | 1, 2 | 39 | |||
AY043269.1 | - | 2, 3 | 307 | |||
BC062950.1 | - | 3, 4 | 666 | 6840162 | ||
CF746424.1 | - | 1, 2 | 143 |