Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnot6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290741.1 | NP_001277670.1 | 10 | 1332 | ||
NM_212484.2 | NP_997649.1 | 11 | 1686 | |||
GenBank mRNA | AK129307.1 | - | 5 | 2624 | ||
AK212064.1 | - | 2 | 170 | |||
AK213936.1 | - | 2 | 159 | |||
AY043269.1 | - | 10 | 1330 | |||
BC023766.1 | - | 6 | 797 | |||
BC037217.1 | - | 1 | 40 | |||
BC049984.1 | - | 6 | 856 | |||
BC057190.1 | - | 5 | 783 | |||
BC062950.1 | - | 11 | 1674 | 6840162 |